Assay Details
| Gene Symbol: Srp68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146032.3 | NP_666144.3 | 6 - 7 | 864 | 100 | |
| GenBank mRNA | AK033010.1 | - | 6 - 7 | 783 | 100 | |
| AK076003.1 | - | 6 - 7 | 761 | 100 | ||
| BC033573.1 | - | 2 - 3 | 164 | 5323201 | 100 | |
| BC070422.1 | - | 6 - 7 | 864 | 6403747 | 100 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC033573.1 transcripts.
Set Membership: |
|

Srp68
Genomic Map