Assay Details
| Gene Symbol: Ctage5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165253.1 | NP_001158725.1 | 2 - 3 | 290 | 78 | |
| NM_001165254.1 | NP_001158726.1 | 2 - 3 | 289 | 78 | ||
| NM_146034.3 | NP_666146.3 | 2 - 3 | 289 | 78 | ||
| GenBank mRNA | AK029466.1 | - | 2 - 3 | 289 | 78 | |
| AK053010.1 | - | 2 - 3 | 223 | 78 | ||
| AK080312.1 | - | 2 - 3 | 265 | 78 | ||
| AK186274.1 | - | 1 - 2 | 83 | 78 | ||
| AK208297.1 | - | 2 - 3 | 162 | 78 | ||
| BC026864.1 | - | 2 - 3 | 241 | 2647214 | 78 | |
| BQ571563.1 | - | 2 - 3 | 289 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctage5
Genomic Map