Assay Details
| Gene Symbol: Mgat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146035.2 | NP_666147.1 | 1 | 406 | 120 | |
| GenBank mRNA | AK083370.1 | - | 1 | 363 | 120 | |
| AK151964.1 | - | 1 | 354 | 120 | ||
| AK152925.1 | - | 1 | 120 | |||
| AK153382.1 | - | 1 | 354 | 120 | ||
| AK166037.1 | - | 1 | 369 | 121 | ||
| AK171000.1 | - | 1 | 409 | 120 | ||
| BC010583.1 | - | 1 | 351 | 3500709 | 120 | |
| BC027169.1 | - | 1 | 351 | 4954047 | 120 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK083370.1,AK151964.1,AK152925.1,AK153382.1,AK171000.1,BC010583.1,BC027169.1,NM_146035.2 transcripts.
Set Membership: |
|

Mgat2
Genomic Map