Assay Details
| Gene Symbol: Ahsa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146036.1 | NP_666148.1 | 1 - 2 | 203 | 89 | |
| GenBank mRNA | AK076069.1 | - | 1 - 2 | 213 | 89 | |
| AK165289.1 | - | 1 - 2 | 223 | 89 | ||
| AK166644.1 | - | 1 - 2 | 176 | 89 | ||
| BC023857.1 | - | 1 - 2 | 170 | 5346745 | 89 | |
| BC025552.1 | - | 1 - 2 | 203 | 5320590 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ahsa1
Genomic Map