Assay Details
| Gene Symbol: Wdr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146039.3 | NP_666151.3 | 12 - 13 | 1907 | 71 | |
| GenBank mRNA | AK052482.1 | - | 12 - 13 | 1871 | 71 | |
| AK077372.1 | - | 2 - 3 | 337 | 71 | ||
| AK144138.1 | - | 12 - 13 | 1907 | 71 | ||
| BC021371.1 | - | 2 - 3 | 150 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr60
Genomic Map