Assay Details
| Gene Symbol: Cdca7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146040.1 | NP_666152.1 | 1 - 2 | 103 | 78 | |
| GenBank mRNA | AK149673.1 | - | 1 - 2 | 81 | 78 | |
| BC006933.1 | - | 1 - 2 | 103 | 3602150 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK149673.1, BC006933.1, NM_146040.1 transcripts.
Set Membership: |
|

Cdca7l
Genomic Map