Assay Details
| Gene Symbol: Lrrc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC019794.1 | - | 1 - 2 | 181 | 4481142 | 77 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC019794.1 transcripts.
Set Membership: |
|

Lrrc3b
Genomic Map