Assay Details
| Gene Symbol: Slc26a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290320.1 | NP_001277249.1 | 10 - 11 | 1565 | 81 | |
| GenBank mRNA | BC027076.1 | - | 10 - 11 | 1565 | 5361175 | 81 |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a8
Genomic Map