Assay Details
| Gene Symbol: Taf6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177798.1 | NP_001171269.1 | 5 - 6 | 652 | 68 | |
| NM_146092.2 | NP_666204.2 | 4 - 5 | 414 | 68 | ||
| GenBank mRNA | AK077149.1 | - | 5 - 6 | 653 | 68 | |
| AK144881.1 | - | 4 - 5 | 381 | 68 | ||
| AK178817.1 | - | 2 - 3 | 186 | 68 | ||
| BC020299.1 | - | 4 - 5 | 387 | 68 | ||
| BC028647.1 | - | 4 - 5 | 361 | 1244243 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taf6l
Genomic Map