Assay Details
| Gene Symbol: Taf6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177798.1 | NP_001171269.1 | 7 - 8 | 850 | 77 | |
| NM_146092.2 | NP_666204.2 | 6 - 7 | 612 | 77 | ||
| GenBank mRNA | AK077149.1 | - | 7 - 8 | 851 | 77 | |
| AK144881.1 | - | 6 - 7 | 579 | 77 | ||
| BC020299.1 | - | 6 - 7 | 585 | 77 | ||
| BC028647.1 | - | 6 - 7 | 559 | 1244243 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taf6l
Genomic Map