Assay Details
| Gene Symbol: Rorb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043354.2 | NP_001036819.1 | 2 - 3 | 730 | 74 | |
| NM_001289921.1 | NP_001276850.1 | 3 - 4 | 249 | 74 | ||
| NM_146095.4 | NP_666207.3 | 2 - 3 | 196 | 74 | ||
| GenBank mRNA | AB258405.1 | - | 2 - 3 | 357 | 74 | |
| AK044421.1 | - | 2 - 3 | 191 | 74 | ||
| AK134790.1 | - | 2 - 3 | 496 | 74 | ||
| BC024768.1 | - | 3 - 4 | 232 | 74 | ||
| BC024842.1 | - | 3 - 4 | 249 | 5358124 | 74 | |
| BC058269.1 | - | 1 - 2 | 103 | 74 | ||
| DQ779923.1 | - | 1 - 2 | 100 | 74 | ||
| DQ779924.1 | - | 2 - 3 | 125 | 74 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC024842.1,NM_001289921.1 transcripts.
Set Membership: |
|

Rorb
Genomic Map