Assay Details
| Gene Symbol: Rorb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043354.2 | NP_001036819.1 | 5 - 6 | 1397 | 80 | |
| NM_001289921.1 | NP_001276850.1 | 6 - 7 | 916 | 80 | ||
| NM_146095.4 | NP_666207.3 | 5 - 6 | 863 | 80 | ||
| GenBank mRNA | AB258405.1 | - | 5 - 6 | 1024 | 80 | |
| AK134790.1 | - | 5 - 6 | 1164 | 80 | ||
| BC024842.1 | - | 6 - 7 | 916 | 5358124 | 80 | |
| BC058269.1 | - | 4 - 5 | 770 | 80 | ||
| DQ779923.1 | - | 4 - 5 | 767 | 80 | ||
| DQ779924.1 | - | 5 - 6 | 792 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rorb
Genomic Map