Assay Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 3 - 4 | 774 | 95 | |
| GenBank mRNA | AK031548.1 | - | 3 - 4 | 774 | 95 | |
| AK040377.1 | - | 2 - 3 | 1005 | 95 | ||
| AK166470.1 | - | 3 - 4 | 904 | 95 | ||
| BC023786.1 | - | 2 - 3 | 152 | 95 | ||
| BC026832.1 | - | 3 - 4 | 824 | 4486176 | 95 | |
| BC036128.1 | - | 3 - 4 | 892 | 4971791 | 95 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK040377.1 transcripts.
Set Membership: |
|

Nrd1
Genomic Map