Assay Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 7 - 8 | 1218 | 81 | |
| GenBank mRNA | AK031548.1 | - | 7 - 8 | 1218 | 81 | |
| AK040377.1 | - | 6 - 7 | 1449 | 81 | ||
| BC023786.1 | - | 6 - 7 | 596 | 81 | ||
| BC026832.1 | - | 7 - 8 | 1268 | 4486176 | 81 | |
| BC036128.1 | - | 7 - 8 | 1336 | 4971791 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrd1
Genomic Map