Assay Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 20 - 21 | 2372 | 110 | |
| GenBank mRNA | AK031548.1 | - | 20 - 21 | 2372 | 110 | |
| AK050944.1 | - | 6 - 7 | 555 | 110 | ||
| AK166470.1 | - | 18 - 19 | 2371 | 110 | ||
| BC023786.1 | - | 19 - 20 | 1750 | 110 | ||
| BC024544.1 | - | 4 - 5 | 357 | 110 | ||
| BC026832.1 | - | 20 - 21 | 2422 | 4486176 | 110 | |
| BC029977.1 | - | 11 - 12 | 1008 | 110 | ||
| BC036128.1 | - | 20 - 21 | 2490 | 4971791 | 110 | |
| U86112.1 | - | 6 - 7 | 620 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nrd1
Genomic Map