Assay Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146150.2 | NP_666262.2 | 24 - 25 | 2832 | 84 | |
| GenBank mRNA | AK031548.1 | - | 24 - 25 | 2832 | 84 | |
| AK050944.1 | - | 10 - 11 | 1015 | 84 | ||
| AK166470.1 | - | 22 - 23 | 2831 | 84 | ||
| BC023786.1 | - | 23 - 24 | 2210 | 84 | ||
| BC024544.1 | - | 8 - 9 | 817 | 84 | ||
| BC026832.1 | - | 24 - 25 | 2882 | 4486176 | 84 | |
| BC029977.1 | - | 15 - 16 | 1468 | 84 | ||
| BC036128.1 | - | 24 - 25 | 2950 | 4971791 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrd1
Genomic Map