Assay Details
| Gene Symbol: Aimp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146165.2 | NP_666277.1 | 1 - 2 | 139 | 70 | |
| GenBank mRNA | BC024480.1 | - | 1 - 2 | 130 | 4977193 | 70 |
| CJ051555.1 | - | 1 - 2 | 131 | 70 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC024480.1,NM_146165.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158561 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158559 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158563 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158558 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158562 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158560 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158557 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993976 | BC026972.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993976 | CX566811.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993976 | NM_001172146.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993976 | NM_146165.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993976 | BC024410.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818469 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499149 | AK016372.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499149 | AK016399.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494234 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 870344 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 69572 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157424 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993976 | BC026972.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993976 | CX566811.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993976 | NM_001172146.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993976 | NM_146165.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993976 | BC024410.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P240020 | BC026972.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P240020 | BY202241.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P240020 | CF732613.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P240020 | CJ051555.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P240020 | NM_001172146.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24158557 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01241916 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157424 | - | - |
| Whole Mouse Genome G4122A | A_52_P240020 | BC026972.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240020 | BY202241.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240020 | CF732613.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240020 | CJ051555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240020 | NM_001172146.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140426 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157424 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04702529 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04702544 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158560 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158559 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158558 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158557 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158561 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158562 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24158563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24158535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04405491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04702543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04405488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04939125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04509505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04509502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04863337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04702538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04509504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04405489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04702546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04939127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04642378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04330768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04702544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24158539 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535507 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535503 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535504 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10535506 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451262_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424151_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535502 | - | - |
| Mouse Genome 430 2.0 Array | 1424151_at | - | - |
| Mouse Genome 430 2.0 Array | 1424151_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451262_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451262_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231872_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231872_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500032696.mm.1 | TC0500003566.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032696.mm.1 | NM_001172146.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032696.mm.1 | NM_001172146.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032696.mm.1 | NM_146165.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032696.mm.1 | NM_146165.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032694.mm.1 | TC0500003566.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032694.mm.1 | NM_001172146.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032694.mm.1 | NM_001172146.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032694.mm.1 | NM_146165.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032694.mm.1 | NM_146165.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032700.mm.1 | TC0500003566.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500032700.mm.1 | NM_001172146.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032700.mm.1 | NM_001172146.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032700.mm.1 | NM_146165.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500032700.mm.1 | NM_146165.2 | Refseq |
| Mouse Expression Set 430 | 1451262_a_at | - | - |
| Mouse Expression Set 430 | 1424151_PM_at | - | - |
| Mouse Expression Set 430 | 1424151_at | - | - |
| Mouse Expression Set 430 | 1451262_PM_a_at | - | - |
| Murine Genome U74v2 Set | 160341_at | - | - |
| Murine Genome U74v2 Set | 160341_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424151_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451262_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451262_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424151_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451262_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451262_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424151_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424151_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 460324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 460324 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000460324 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244330 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244330 | - | - |
| mouse-6 v1.1 (Illumina) | scl25798.3_114-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 460324 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25798.3_114-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 460324 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1244330 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2943217 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244330 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2943217 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl25798.3_114-S | - | - |

Aimp2
Genomic Map