Assay Details
| Gene Symbol: Tspan33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146173.3 | NP_666285.1 | 2 - 3 | 389 | 61 | |
| GenBank mRNA | AF276890.1 | - | 2 - 3 | 388 | 61 | |
| AK004954.1 | - | 2 - 3 | 363 | 61 | ||
| AK162631.1 | - | 1 - 2 | 1984 | 61 | ||
| BC024685.1 | - | 2 - 3 | 347 | 3981492 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tspan33
Genomic Map