| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700000641.mm.1 |
TC0700000056.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000641.mm.1 |
NM_146178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000641.mm.1 |
NM_146178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000649.mm.1 |
TC0700000056.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000649.mm.1 |
NM_146178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000649.mm.1 |
NM_146178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000645.mm.1 |
TC0700000056.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000645.mm.1 |
NM_146178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000645.mm.1 |
NM_146178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000644.mm.1 |
TC0700000056.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000644.mm.1 |
NM_146178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000644.mm.1 |
NM_146178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000646.mm.1 |
TC0700000056.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000646.mm.1 |
NM_146178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000646.mm.1 |
NM_146178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000647.mm.1 |
TC0700000056.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000647.mm.1 |
NM_146178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000647.mm.1 |
NM_146178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000639.mm.1 |
TC0700000056.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000639.mm.1 |
NM_146178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000639.mm.1 |
NM_146178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000642.mm.1 |
TC0700000056.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000642.mm.1 |
NM_146178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000642.mm.1 |
NM_146178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000643.mm.1 |
TC0700000056.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000643.mm.1 |
NM_146178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000643.mm.1 |
NM_146178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000648.mm.1 |
TC0700000056.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700000648.mm.1 |
NM_146178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700000648.mm.1 |
NM_146178.2 |
Refseq |