Assay Details
| Gene Symbol: Picalm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252520.1 | NP_001239449.1 | 19 - 20 | 2191 | 70 | |
| NM_001252521.1 | NP_001239450.1 | 18 - 19 | 2182 | 70 | ||
| NM_001252522.1 | NP_001239451.1 | 18 - 19 | 2056 | 70 | ||
| NM_001252523.1 | NP_001239452.1 | 18 - 19 | 2041 | 70 | ||
| NM_001252524.1 | NP_001239453.1 | 17 - 18 | 2017 | 70 | ||
| NM_146194.4 | NP_666306.2 | 19 - 20 | 2206 | 70 | ||
| GenBank mRNA | AK085472.2 | - | 14 - 15 | 1251 | 70 | |
| AK144960.1 | - | 19 - 20 | 1947 | 70 | ||
| AK162360.1 | - | 19 - 20 | 1884 | 70 | ||
| AK165066.1 | - | 13 - 14 | 1153 | 70 | ||
| AK170373.1 | - | 17 - 18 | 2037 | 70 | ||
| AK220291.1 | - | 19 - 20 | 1907 | 70 | ||
| AY206701.1 | - | 19 - 20 | 1890 | 70 | ||
| BC011470.1 | - | 19 - 20 | 1901 | 2651109 | 70 | |
| BC021491.1 | - | 17 - 18 | 1966 | 5345558 | 70 | |
| BC023843.1 | - | 18 - 19 | 1930 | 5346041 | 70 | |
| BC025566.1 | - | 18 - 19 | 2079 | 5322606 | 70 | |
| BC027116.1 | - | 15 - 16 | 1478 | 70 | ||
| BC057683.1 | - | 18 - 19 | 1992 | 3671962 | 70 | |
More Information
Set Membership: |
|

Picalm
Genomic Map