Assay Details
| Gene Symbol: Acsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177978.1 | NP_001171449.1 | 1 - 2 | 100 | 109 | |
| NM_146197.4 | NP_666309.1 | 1 - 2 | 100 | 109 | ||
| GenBank mRNA | AK057650.1 | - | 1 - 2 | 100 | 109 | |
| AK143757.1 | - | 1 - 2 | 96 | 109 | ||
| AK165353.1 | - | 1 - 2 | 98 | 109 | ||
| AK165401.1 | - | 1 - 2 | 89 | 109 | ||
| BC031140.1 | - | 1 - 2 | 76 | 4972673 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK057650.1, AK143757.1, AK165353.1, AK165401.1, BC031140.1, NM_001177978.1, NM_146197.4 transcripts.
Set Membership: |
|

Acsm2
Genomic Map