Assay Details
| Gene Symbol: Stard13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146258.2 | NP_666370.3 | 6 - 7 | 2022 | 96 | |
| GenBank mRNA | BC027830.1 | - | 6 - 7 | 2034 | 5320966 | 96 |
| DV651942.1 | - | 2 - 3 | 281 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stard13
Genomic Map