Assay Details
| Gene Symbol: Olfr1394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146276.1 | NP_666388.1 | 1 | 727 | 132 | |
| GenBank mRNA | BC127968.1 | - | 1 | 731 | 40135034 | 132 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113954 | NM_146276.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2113954 | BC127968.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236880 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165518 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113954 | NM_146276.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2113954 | BC127968.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285462 | NM_146276.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285462 | BC127968.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27781570 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165518 | - | - |
| Whole Mouse Genome G4122A | A_51_P285462 | NM_146276.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P285462 | BC127968.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104394 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165518 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09294454 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09294452 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09016618 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09294449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27781575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09705829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09294441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09690400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09015476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09243218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09618918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09294443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09243220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09083844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09035304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09035307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09015473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09106606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09294449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27781560 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10375528 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10375527 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 530624 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 360095 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 360095 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000360095 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2616214 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2616214 | - | - |
| mouse-6 v1.1 (Illumina) | scl14141.1.1_218-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 360095 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl14141.1.1_218-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 360095 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2616214 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2616214 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl14141.1.1_218-S | - | - |

Olfr1394
Genomic Map