Assay Details
| Gene Symbol: Olfr1491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146345.1 | NP_666457.1 | 1 | 911 | 112 | |
| GenBank mRNA | BC128268.1 | - | 1 | 911 | 40043876 | 112 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233846 | BC128268.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233846 | NM_146345.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233846 | BC128268.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P233846 | NM_146345.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413320 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121505 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233846 | BC128268.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233846 | NM_146345.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233846 | BC128268.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P233846 | NM_146345.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233846 | BC128268.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233846 | NM_146345.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233846 | BC128268.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233846 | NM_146345.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31965411 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121505 | - | - |
| Whole Mouse Genome G4122A | A_51_P233846 | BC128268.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233846 | NM_146345.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233846 | BC128268.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P233846 | NM_146345.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136510 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121505 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14208715 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14155622 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14155612 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14155610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31965438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14410398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14149836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14155614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14281330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14299157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14299151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14192249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31965449 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10461825 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461824 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900003046.mm.1 | NM_146345.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003046.mm.1 | NM_146345.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003046.mm.1 | TC1900000281.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 353171 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 730113 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000730113 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1256579 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256579 | - | - |
| mouse-6 v1.1 (Illumina) | scl6280.1.1_66-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 730113 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl6280.1.1_66-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 730113 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1256579 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256579 | - | - |

Olfr1491
Genomic Map