Assay Details
| Gene Symbol: Olfr368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146374.1 | NP_666486.1 | 1 | 935 | 107 | |
| GenBank mRNA | BC104308.1 | - | 1 | 935 | 40044244 | 107 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974917 | NM_146374.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974917 | BC104308.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974917 | BC104309.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164512 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974917 | NM_146374.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974917 | BC104308.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974917 | BC104309.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411801 | NM_146374.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411801 | BC104308.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P411801 | BC104309.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21141514 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164512 | - | - |
| Whole Mouse Genome G4122A | A_51_P411801 | NM_146374.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P411801 | BC104308.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P411801 | BC104309.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118656 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164512 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01168315 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01168288 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01168295 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01168293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21141553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01726707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02087185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02101432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141522 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01168296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02101434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01168288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01778287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01168297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141517 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01168310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21141544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01168300 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10471783 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10471782 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 873620 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2230010 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002230010 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218591 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218591 | - | - |
| mouse-6 v1.1 (Illumina) | scl5045.1.1_49-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2230010 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl5045.1.1_49-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2230010 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1218591 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1218591 | - | - |

Olfr368
Genomic Map