Assay Details
Gene Symbol: Olfr1026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146584.2 | NP_666795.2 | 1 | 740 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102563 | NM_146584.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134991 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102563 | NM_146584.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P478498 | NM_146584.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399973 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399960 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399975 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399964 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399957 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399961 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399982 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399955 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399985 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399947 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399966 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399959 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21400002 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399962 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399969 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399971 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399958 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399999 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399984 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399978 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399986 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399977 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399988 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399965 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399952 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21399990 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134991 | - | - |
Whole Mouse Genome G4122A | A_51_P478498 | NM_146584.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119836 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134991 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02103065 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01502223 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02103068 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01502229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21400032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21399985 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01906811 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399936 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399941 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400023 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399966 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400016 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02186238 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400002 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399977 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400019 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399947 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399952 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399988 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399968 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399959 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399955 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399958 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400027 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399971 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400005 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399945 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399969 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399999 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01675956 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399975 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399960 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400026 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01906813 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399957 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400008 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399990 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399978 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399973 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399961 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01502222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399992 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399985 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399938 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399994 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399934 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21400024 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399986 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21399950 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10473480 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10473479 | - | - |
Affymetrix Mouse Clariom S Array | PSR0200009967.mm.1 | NM_146584.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009967.mm.1 | NM_146584.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009967.mm.1 | TC0200001256.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 825969 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1340397 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001340397 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2641178 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2641178 | - | - |
mouse-6 v1.1 (Illumina) | scl4934.1.1_214-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1340397 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl4934.1.1_214-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340397 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2641178 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2641178 | - | - |
