Assay Details
| Gene Symbol: Olfr700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146600.1 | NP_666811.1 | 1 | 460 | 76 | |
| GenBank mRNA | BC104090.1 | - | 1 | 460 | 76 | |
| BC104091.2 | - | 1 | 460 | 40042950 | 76 | |
| BC106811.1 | - | 1 | 460 | 76 | ||
| BC106812.2 | - | 1 | 460 | 40043224 | 76 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021953 | BC106811.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021953 | BC106812.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021953 | BC104091.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021953 | NM_146600.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021953 | BC104090.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021953 | BC106811.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021953 | BC106812.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021953 | BC104091.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021953 | NM_146600.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021953 | BC104090.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P434227 | BC106811.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P434227 | BC106812.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P434227 | BC104091.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P434227 | NM_146600.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P434227 | BC104090.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25396641 | - | - |
| Whole Mouse Genome G4122A | A_51_P434227 | BC106811.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P434227 | BC106812.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P434227 | BC104091.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P434227 | NM_146600.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P434227 | BC104090.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129688 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109274 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06280327 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06280334 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06280318 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06542555 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06167754 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06542570 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06280304 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06542567 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06167753 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06542573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25396661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06167745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25396641 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566602 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566601 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 321124 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3800576 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003800576 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219081 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219081 | - | - |
| mouse-6 v1.1 (Illumina) | scl30860.1.1_80-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3800576 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30860.1.1_80-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800576 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219081 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219081 | - | - |

Olfr700
Genomic Map