| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900002968.mm.1 |
NM_146698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002968.mm.1 |
NM_146698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002968.mm.1 |
TC1900000253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002970.mm.1 |
NM_146698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002970.mm.1 |
NM_146698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002970.mm.1 |
TC1900000253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900002969.mm.1 |
NM_146698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002969.mm.1 |
NM_146698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900002969.mm.1 |
TC1900000253.mm.1 |
MTA TC |