Assay Details
| Gene Symbol: Olfr248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146714.2 | NP_666925.2 | 1 | 571 | 118 | |
| GenBank mRNA | BC131960.1 | - | 1 | 571 | 40130237 | 118 |
| BC131962.1 | - | 1 | 571 | 40130239 | 118 | |
| Gene Symbol: Olfr417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207137.3 | NP_997020.3 | 1 | 546 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122211 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1952038 | NM_207137.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150209 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159936 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122211 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1952038 | NM_207137.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128056 | NM_146714.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128056 | BC131960.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128056 | BC131962.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128056 | NM_207137.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20842059 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150209 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159936 | - | - |
| Whole Mouse Genome G4122A | A_51_P128056 | NM_146714.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128056 | BC131960.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128056 | BC131962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128056 | NM_207137.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108293 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122211 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134211 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159936 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150209 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00733267 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00150554 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00150563 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00733276 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00150559 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00733270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20842127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00077670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00967347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00621405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00077671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00265195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00225282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00278197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00077666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00259414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00212889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00967346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00780506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00577148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00061818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00278203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00331538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00739548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00077678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20842103 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351962 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351965 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351963 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351961 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100013687.mm.1 | NM_207137.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013687.mm.1 | NM_207137.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013687.mm.1 | TC0100001647.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 897565 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5720647 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720647 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2773061 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2773061 | - | - |
| mouse-6 v1.1 (Illumina) | scl14315.1.1_235-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720647 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl14315.1.1_235-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720647 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2846636 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2970309 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2970309 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2846636 | - | - |

Olfr248
Genomic Map