Assay Details
| Gene Symbol: Olfr421-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146720.3 | NP_666931.3 | 1 | 85 | 69 | |
| NR_047667.1 | 1 | 85 | 69 | |||
| GenBank mRNA | BC119233.1 | - | 1 | 85 | 8733982 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969037 | NM_146720.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969037 | BC119233.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117656 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162553 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157032 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969037 | NM_146720.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969037 | BC119233.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P113321 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117656 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157032 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162553 | - | - |
| Whole Mouse Genome G4122A | A_51_P113321 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129601 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117656 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157032 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20841187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00910575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00962123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00145139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20841122 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351901 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351900 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351902 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351899 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100013621.mm.1 | NR_047667.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013621.mm.1 | TC0100001642.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4810463 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004810463 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2771458 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2771458 | - | - |
| mouse-6 v1.1 (Illumina) | scl14317.1.1_213-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4810463 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl14317.1.1_213-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4810463 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2771458 | - | - |

Olfr421-ps1
Genomic Map