Assay Details
| Gene Symbol: Olfr1362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146744.2 | NP_666955.2 | 1 | 476 | 112 | |
| GenBank mRNA | BC119336.1 | - | 1 | 476 | 8734085 | 112 |
| BC119338.1 | - | 1 | 476 | 8734087 | 112 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018691 | BC119338.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018691 | NM_146744.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018691 | BC119336.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176064 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018691 | BC119338.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018691 | NM_146744.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018691 | BC119336.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186896 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P150242 | BC119338.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P150242 | NM_146744.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P150242 | BC119336.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28873747 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176064 | - | - |
| Whole Mouse Genome G4122A | A_51_P150242 | BC119338.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P150242 | NM_146744.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P150242 | BC119336.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133989 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176064 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10844603 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10844613 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10637976 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10637986 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10844614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28873735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10632744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10844611 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10637976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10637981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10844599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10844602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10637978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10827761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28873747 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408063 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408062 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 817488 | - | - |
| ABI Mouse Genome Survey Microarray | 899511 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1230184 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1230184 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001230184 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2751176 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2751176 | - | - |
| mouse-6 v1.1 (Illumina) | scl11534.1.1_37-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1230184 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl11534.1.1_37-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1230184 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2751183 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2751183 | - | - |

Olfr1362
Genomic Map