Assay Details
Gene Symbol: Olfr975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146828.1 | NP_667039.1 | 1 | 152 | 87 | |
GenBank mRNA | BC100677.1 | - | 1 | 151 | 87 | |
BC101947.1 | - | 1 | 151 | 87 | ||
BC141889.1 | - | 1 | 150 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081975 | BC101947.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081975 | BC141889.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081975 | NM_146828.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2081975 | BC100677.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P116028 | BC101947.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P116028 | BC141889.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P116028 | NM_146828.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P116028 | BC100677.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148709 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081975 | BC101947.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081975 | BC141889.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081975 | NM_146828.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2081975 | BC100677.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P116028 | BC101947.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P116028 | BC141889.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P116028 | NM_146828.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P116028 | BC100677.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234985 | BC101947.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234985 | BC141889.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234985 | NM_146828.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P234985 | BC100677.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440664 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440637 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440662 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440643 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440655 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440656 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440673 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440652 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440677 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440671 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440657 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440667 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440649 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440645 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440659 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440642 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440650 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440644 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440675 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440638 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440647 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440640 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440666 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440663 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26440668 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148709 | - | - |
Whole Mouse Genome G4122A | A_51_P234985 | BC101947.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P234985 | BC141889.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P234985 | NM_146828.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P234985 | BC100677.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116028 | BC101947.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116028 | BC141889.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116028 | NM_146828.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116028 | BC100677.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148709 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07827540 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07827542 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08149461 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07827550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440712 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26440662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440677 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440652 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07574413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440633 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440632 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440643 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440658 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440662 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07827541 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440673 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440634 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440668 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440650 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440630 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440637 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440638 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440675 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440647 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440648 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440667 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08149463 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440644 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440642 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26440663 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10592456 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10592455 | - | - |
Affymetrix Mouse Clariom S Array | PSR0900017261.mm.1 | NM_146828.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900017261.mm.1 | NM_146828.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900017261.mm.1 | TC0900002087.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 441140 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1660347 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001660347 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2625468 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2625468 | - | - |
mouse-6 v1.1 (Illumina) | scl6610.1.1_106-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1660347 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl6610.1.1_106-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660347 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2625468 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2625468 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl6610.1.1_106-S | - | - |
