Assay Details
| Gene Symbol: Olfr272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146839.1 | NP_667050.1 | 1 | 456 | 56 | |
| GenBank mRNA | BC128006.1 | - | 1 | 460 | 40135971 | 56 |
More Information
Updated Mapping Information
Set Membership: |
|

Olfr272
Genomic Map