Assay Details
| Gene Symbol: Olfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146921.2 | NP_667132.1 | 3 | 970 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
G-protein coupled receptor signaling pathway
sensory perception of smell
response to stimulus
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
olfactory receptor activity
signal transduction
G-protein coupled receptor signaling pathway
sensory perception of smell
response to stimulus
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
olfactory receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2053384 | NM_146921.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106193 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053384 | NM_146921.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P413204 | S73846.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P348922 | NM_146921.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27916956 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106193 | - | - |
| Whole Mouse Genome G4122A | A_51_P348922 | NM_146921.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P413204 | S73846.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133560 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106193 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09243893 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09243897 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09243910 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09243912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27916930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09407501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09407507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09612442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09035850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09407502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09407503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09243913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09436380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09243908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27916942 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10388266 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10388265 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100029323.mm.1 | NM_146921.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100029323.mm.1 | NM_146921.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100029323.mm.1 | TC1100003165.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 372211 | - | - |
| ABI Mouse Genome Survey Microarray | 864063 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3130333 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003130333 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255360 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255360 | - | - |
| mouse-6 v1.1 (Illumina) | scl00258923.1_45-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3130333 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00258923.1_45-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3130333 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1255360 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255360 | - | - |

Olfr1
Genomic Map