Assay Details
Gene Symbol: Olfr352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146940.1 | NP_667151.1 | 1 | 534 | 74 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046273 | NM_146940.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179906 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046273 | NM_146940.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P308778 | NM_146940.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140323 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140325 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140319 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140328 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140321 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140324 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21140315 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179906 | - | - |
Whole Mouse Genome G4122A | A_51_P308778 | NM_146940.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113420 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21140294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140338 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01231349 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02170372 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01231370 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01231355 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01778145 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01908702 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140309 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01711548 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140300 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140324 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140332 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01231352 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01778147 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140315 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01852263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01231367 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01231353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140319 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140323 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21140329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02204215 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10471762 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10471761 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 321999 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5570452 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005570452 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2746970 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2746970 | - | - |
mouse-6 v1.1 (Illumina) | scl5049.1.1_37-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5570452 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl5049.1.1_37-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570452 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2809136 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2809136 | - | - |
