Assay Details
Gene Symbol: Olfr171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146958.2 | NP_667169.2 | 1 | 262 | 82 |
Gene Symbol: Olfr169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011855.1 | NP_001011855.1 | 1 | 259 | 82 |
Gene Symbol: Olfr170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146957.1 | NP_667168.1 | 1 | 259 | 82 | |
GenBank mRNA | BC132585.1 | - | 1 | 296 | 40130862 | 82 |
BC132587.1 | - | 1 | 296 | 40130864 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P237973 | NM_146957.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P237973 | BC132585.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P237973 | BC132587.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P237973 | NM_146957.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P237973 | BC132585.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P237973 | BC132587.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P26086 | NM_146958.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P26086 | NM_146958.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P123148 | NM_001011855.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P237973 | NM_146957.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P237973 | BC132585.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P237973 | BC132587.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P237973 | NM_146957.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P237973 | BC132585.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P237973 | BC132587.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P123148 | NM_001011855.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P26086 | NM_146958.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P26086 | NM_146958.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P237973 | NM_146957.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P237973 | BC132585.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P237973 | BC132587.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P237973 | NM_146957.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P237973 | BC132585.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P237973 | BC132587.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P26086 | NM_146958.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P26086 | NM_146958.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584483 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584297 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584295 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584474 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584462 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584307 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584453 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584490 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584464 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584469 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584477 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584436 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584476 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584461 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584317 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584480 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584322 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584471 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584410 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584479 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584493 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584393 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584468 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584467 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584455 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584457 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584492 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584440 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584487 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584406 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584298 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584458 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584451 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584456 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584387 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584481 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584452 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584463 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584482 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584296 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584473 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584374 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30584394 | - | - |
Whole Mouse Genome G4122A | A_52_P26086 | NM_146958.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P26086 | NM_146958.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P237973 | NM_146957.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P237973 | BC132585.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P237973 | BC132587.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P237973 | NM_146957.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P237973 | BC132585.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P237973 | BC132587.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128572 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130316 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12939070 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12990522 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12884309 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12862744 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12789922 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12700271 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12939074 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12789918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584480 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30584471 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584438 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12990530 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584501 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12952670 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584479 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584322 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584480 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584477 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12731156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584482 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584408 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584467 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584458 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584459 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584369 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12990521 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12882826 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584451 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12789919 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584496 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12765288 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12531720 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584374 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12700271 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584488 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584453 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12531721 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584469 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584471 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584483 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584387 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584481 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12705425 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584464 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584432 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584433 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584461 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12952669 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584436 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584455 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584298 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584456 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13047709 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584410 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584366 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13047700 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584406 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584473 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584310 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584492 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584493 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584288 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584440 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584393 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584452 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12783738 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584476 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12884309 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584462 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584399 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584468 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584293 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584503 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30584319 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10438434 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10438432 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10438430 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10438430 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10438433 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10438429 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10438431 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10438429 | - | - |
Affymetrix Mouse Clariom S Array | PSR1600010988.mm.1 | NM_146958.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600010988.mm.1 | NM_146958.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600010988.mm.1 | TC1600001333.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600010986.mm.1 | NM_146957.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600010986.mm.1 | NM_146957.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600010986.mm.1 | TC1600001332.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600010985.mm.1 | TC1600001332.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600010987.mm.1 | TC1600001332.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600010984.mm.1 | NM_001011855.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600010984.mm.1 | NM_001011855.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600010984.mm.1 | TC1600001331.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 436014 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6450605 | - | - |
MouseWG-6 v2 Expression BeadChip | 1340139 | - | - |
MouseWG-6 v2 Expression BeadChip | 5550110 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6450605 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5550110 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1340139 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001340139 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005550110 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006450605 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2695874 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2778293 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2721856 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2778293 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2695874 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2721856 | - | - |
mouse-6 v1.1 (Illumina) | scl00258959.1_326-S | - | - |
mouse-6 v1.1 (Illumina) | scl00258960.1_326-S | - | - |
mouse-6 v1.1 (Illumina) | scl0258960.2_9-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5550110 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6450605 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1340139 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258960.2_9-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00258960.1_326-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00258959.1_326-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550110 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6450605 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340139 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2778293 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2695874 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2721856 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2778293 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2695874 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2721856 | - | - |
