Assay Details
Gene Symbol: Olfr1270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146985.2 | NP_667196.1 | 1 | 680 | 68 | |
GenBank mRNA | BC119113.1 | - | 1 | 680 | 8733862 | 68 |
BC119115.1 | - | 1 | 680 | 8733864 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181490 | NM_146985.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181490 | BC119113.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2181490 | BC119115.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128225 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181490 | NM_146985.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181490 | BC119113.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2181490 | BC119115.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167198 | NM_146985.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167198 | BC119113.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167198 | BC119115.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128225 | - | - |
Whole Mouse Genome G4122A | A_51_P167198 | NM_146985.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P167198 | BC119113.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P167198 | BC119115.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102314 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125410 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21416011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21415991 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416070 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416055 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416083 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416061 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01473885 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416091 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416078 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416066 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416056 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416059 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416069 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416074 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416086 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416068 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416081 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416089 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416085 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416093 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416076 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21416088 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10484874 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10484873 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484872 | - | - |
Affymetrix Mouse Clariom S Array | PSR0200034136.mm.1 | NM_146985.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034136.mm.1 | NM_146985.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034136.mm.1 | TC0200004082.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 330600 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2120601 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 100050128 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002120601 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2639417 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2639417 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2535092 | - | - |
mouse-6 v1.1 (Illumina) | GI_22128966-S | - | - |
mouse-6 v1.1 (Illumina) | scl4262.1.1_34-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2120601 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 100050128 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl4262.1.1_34-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2120601 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100050128 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2639417 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2639417 | - | - |
