Assay Details
| Gene Symbol: Olfr1054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147019.1 | NP_667230.1 | 1 | 849 | 122 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1974872 | NM_147019.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731784 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117754 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1974872 | NM_147019.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P201302 | NM_147019.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117754 | - | - |
| Whole Mouse Genome G4122A | A_51_P201302 | NM_147019.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114113 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21401823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01740653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01740647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01740648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401822 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01237675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01740661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01740658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01511358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01740660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01237680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21401784 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484587 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484586 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 655604 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1400121 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1400121 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001400121 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2637912 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2637912 | - | - |
| mouse-6 v1.1 (Illumina) | scl4340.1.1_82-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1400121 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl4340.1.1_82-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1400121 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2637912 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2637912 | - | - |

Olfr1054
Genomic Map