| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200022083.mm.1 |
NM_001077510.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022083.mm.1 |
NM_201616.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022083.mm.1 |
TC0200002700.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200022093.mm.1 |
NM_001077507.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022093.mm.1 |
NM_001077510.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022093.mm.1 |
NM_010309.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022093.mm.1 |
NM_019690.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022093.mm.1 |
NM_022000.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022093.mm.1 |
NM_201616.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022093.mm.1 |
NM_201618.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022093.mm.1 |
NR_003258.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022093.mm.1 |
NR_003258.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022093.mm.1 |
TC0200002700.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200022082.mm.1 |
NM_001077510.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022082.mm.1 |
NM_201616.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200022082.mm.1 |
TC0200002700.mm.1 |
MTA TC |