Assay Details
| Gene Symbol: Abi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077192.2 | NP_001070660.1 | 6 - 7 | 985 | 94 | |
| NM_007380.3 | NP_031406.2 | 6 - 7 | 985 | 94 | ||
| GenBank mRNA | AF420251.1 | - | 6 - 7 | 921 | 94 | |
| AK151363.1 | - | 6 - 7 | 964 | 94 | ||
| AK153130.1 | - | 6 - 7 | 893 | 94 | ||
| AK164040.1 | - | 6 - 7 | 901 | 94 | ||
| AK165924.1 | - | 6 - 7 | 958 | 94 | ||
| AK167328.1 | - | 6 - 7 | 944 | 94 | ||
| AK167744.1 | - | 6 - 7 | 932 | 94 | ||
| AY033645.1 | - | 6 - 7 | 829 | 94 | ||
| U17698.1 | - | 5 - 6 | 611 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abi1
Genomic Map