Assay Details
| Gene Symbol: Apoc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007385.3 | NP_031411.1 | 1 - 2 | 158 | 61 | |
| GenBank mRNA | BB872041.1 | - | 1 - 2 | 156 | 61 | |
| BC024657.1 | - | 1 - 2 | 114 | 4196217 | 61 | |
| BC100593.1 | - | 1 - 2 | 158 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460332 | NM_007385.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460332 | BC024657.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460332 | NM_007385.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P460332 | BC024657.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366772 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886825 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154921 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460332 | NM_007385.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460332 | BC024657.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460332 | NM_007385.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P460332 | BC024657.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24994694 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154921 | - | - |
| Whole Mouse Genome G4122A | A_51_P460332 | NM_007385.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460332 | BC024657.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460332 | NM_007385.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460332 | BC024657.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137667 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154921 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06453916 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06453915 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06372917 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06453917 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06372918 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06372921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24994639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06507337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05896361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06453917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06372923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24994694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06596783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06372916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06381782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05896362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06236102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06453918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06042780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24994728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24994719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24994697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24994716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06372922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06347177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06372918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06618270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06507341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06372924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06596781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06453916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24994724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24994725 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10560616 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560617 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560615 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418708_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10560614 | - | - |
| Mouse Genome 430 2.0 Array | 1418708_at | - | - |
| Mouse Genome 430 2.0 Array | 1418708_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11425_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11425_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700022012.mm.1 | TC0700002495.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022012.mm.1 | NM_007385.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022016.mm.1 | TC0700002495.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022016.mm.1 | NM_007385.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022013.mm.1 | TC0700002495.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022013.mm.1 | NM_007385.3 | Refseq |
| Mouse Expression Set 430 | 1418708_at | - | - |
| Mouse Expression Set 430 | 1418708_PM_at | - | - |
| Murine Genome U74v2 Set | 101463_PM_at | - | - |
| Murine Genome U74v2 Set | 101463_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418708_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418708_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418708_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418708_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 488439 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1580039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001580039 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234764 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234764 | - | - |
| mouse-6 v1.1 (Illumina) | scl011425.3_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1580039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl011425.3_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1580039 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3009225 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3009225 | - | - |

Apoc4
Genomic Map