Assay Details
| Gene Symbol: Adam8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089086.1 | - | 12 - 13 | 1284 | 73 | |
| AK089169.1 | - | 12 - 13 | 1306 | 73 | ||
| AK150411.1 | - | 12 - 13 | 1286 | 73 | ||
| AK151535.1 | - | 4 - 5 | 515 | 73 | ||
| AK152675.1 | - | 12 - 13 | 1303 | 73 | ||
| AK152769.1 | - | 12 - 13 | 1285 | 73 | ||
| AK153065.1 | - | 12 - 13 | 1285 | 73 | ||
| AK155772.1 | - | 12 - 13 | 1284 | 73 | ||
| AK155912.1 | - | 12 - 13 | 1304 | 73 | ||
| AK160041.1 | - | 12 - 13 | 1287 | 73 | ||
| AK170019.1 | - | 12 - 13 | 1309 | 73 | ||
| AK171060.1 | - | 12 - 13 | 1284 | 73 | ||
| BC025584.1 | - | 12 - 13 | 1338 | 5324943 | 73 | |
| X13335.1 | - | 12 - 13 | 1283 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adam8
Genomic Map