Assay Details
| Gene Symbol: Adam8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089086.1 | - | 20 - 21 | 2256 | 70 | |
| AK134118.1 | - | 9 - 10 | 2303 | 70 | ||
| AK150411.1 | - | 20 - 21 | 2258 | 70 | ||
| AK151535.1 | - | 12 - 13 | 1487 | 70 | ||
| AK152675.1 | - | 20 - 21 | 2275 | 70 | ||
| AK152769.1 | - | 20 - 21 | 2257 | 70 | ||
| AK153065.1 | - | 20 - 21 | 2257 | 70 | ||
| AK155772.1 | - | 20 - 21 | 2256 | 70 | ||
| AK155912.1 | - | 20 - 21 | 2276 | 70 | ||
| AK160041.1 | - | 20 - 21 | 2259 | 70 | ||
| AK170019.1 | - | 20 - 21 | 2281 | 70 | ||
| AK171022.1 | - | 18 - 19 | 2833 | 70 | ||
| AK171060.1 | - | 20 - 21 | 2256 | 70 | ||
| BC025584.1 | - | 20 - 21 | 2310 | 5324943 | 70 | |
| X13335.1 | - | 20 - 21 | 2255 | 70 | ||
More Information
Set Membership: |
|

Adam8
Genomic Map