Assay Details
| Gene Symbol: Ager | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271422.1 | NP_001258351.1 | 2 - 3 | 201 | 58 | |
| NM_007425.3 | NP_031451.2 | 2 - 3 | 201 | 58 | ||
| NR_073173.1 | 2 - 3 | 201 | 58 | |||
| NR_073175.1 | 1 - 2 | 396 | 58 | |||
| NR_073176.1 | 1 - 2 | 396 | 58 | |||
| GenBank mRNA | AB207883.1 | - | 2 - 3 | 161 | 58 | |
| AK004802.1 | - | 2 - 3 | 189 | 58 | ||
| BC061182.1 | - | 2 - 3 | 189 | 30300427 | 58 | |
| EU520325.1 | - | 2 - 3 | 157 | 58 | ||
| EU570240.1 | - | 2 - 3 | 157 | 58 | ||
| EU570241.1 | - | 1 - 2 | 352 | 58 | ||
| EU570242.1 | - | 2 - 3 | 157 | 58 | ||
| EU570243.1 | - | 2 - 3 | 157 | 58 | ||
| EU570244.1 | - | 1 - 2 | 352 | 58 | ||
| EU570245.1 | - | 1 - 2 | 352 | 58 | ||
| EU570246.1 | - | 2 - 3 | 157 | 58 | ||
| EU570247.1 | - | 1 - 2 | 352 | 58 | ||
| EU906858.1 | - | 2 - 3 | 157 | 58 | ||
| EU906859.1 | - | 2 - 3 | 157 | 58 | ||
| EU906862.1 | - | 2 - 3 | 157 | 58 | ||
| EU906864.1 | - | 1 - 2 | 352 | 58 | ||
| GU164728.1 | - | 2 - 3 | 176 | 58 | ||
| GU164729.1 | - | 2 - 3 | 176 | 58 | ||
| GU164730.1 | - | 2 - 3 | 176 | 58 | ||
| GU164731.1 | - | 2 - 3 | 176 | 58 | ||
| GU164732.1 | - | 2 - 3 | 176 | 58 | ||
| GU164734.1 | - | 2 - 3 | 177 | 58 | ||
| KC692918.1 | - | 2 - 3 | 157 | 58 | ||
| L33412.1 | - | 2 - 3 | 162 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ager
Genomic Map