Assay Details
| Gene Symbol: Ager | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271422.1 | NP_001258351.1 | 6 - 7 | 728 | 82 | |
| NM_001271423.1 | NP_001258352.1 | 6 - 7 | 701 | 82 | ||
| NM_001271424.1 | NP_001258353.1 | 5 - 6 | 823 | 82 | ||
| NM_007425.3 | NP_031451.2 | 6 - 7 | 728 | 82 | ||
| NR_073173.1 | 6 - 7 | 728 | 82 | |||
| NR_073175.1 | 5 - 6 | 923 | 82 | |||
| NR_073176.1 | 5 - 6 | 923 | 82 | |||
| GenBank mRNA | AB207883.1 | - | 6 - 7 | 691 | 82 | |
| AK004802.1 | - | 6 - 7 | 717 | 83 | ||
| BC061182.1 | - | 6 - 7 | 716 | 30300427 | 82 | |
| EU520325.1 | - | 6 - 7 | 684 | 82 | ||
| EU570240.1 | - | 6 - 7 | 684 | 82 | ||
| EU570241.1 | - | 5 - 6 | 879 | 82 | ||
| EU570242.1 | - | 6 - 7 | 684 | 82 | ||
| EU570243.1 | - | 6 - 7 | 684 | 82 | ||
| EU570244.1 | - | 5 - 6 | 879 | 82 | ||
| EU570245.1 | - | 5 - 6 | 879 | 82 | ||
| EU570246.1 | - | 5 - 6 | 841 | 82 | ||
| EU570247.1 | - | 5 - 6 | 879 | 82 | ||
| EU906863.1 | - | 6 - 7 | 657 | 82 | ||
| EU906864.1 | - | 5 - 6 | 879 | 82 | ||
| GU164727.1 | - | 5 - 6 | 798 | 82 | ||
| GU164728.1 | - | 6 - 7 | 703 | 82 | ||
| GU164730.1 | - | 6 - 7 | 703 | 82 | ||
| GU164731.1 | - | 6 - 7 | 706 | 82 | ||
| KC692918.1 | - | 6 - 7 | 684 | 82 | ||
| L33412.1 | - | 6 - 7 | 692 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ager
Genomic Map