Assay Details
| Gene Symbol: Ager | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007425.3 | NP_031451.2 | 9 - 10 | 1035 | 97 | |
| NR_073175.1 | 8 - 9 | 1230 | 97 | |||
| GenBank mRNA | BC061182.1 | - | 9 - 10 | 1023 | 30300427 | 97 |
| EU520325.1 | - | 9 - 10 | 991 | 97 | ||
| EU570241.1 | - | 8 - 9 | 1186 | 97 | ||
| EU906857.1 | - | 4 - 5 | 231 | 97 | ||
| GU164728.1 | - | 10 - 11 | 931 | 97 | ||
| GU164737.1 | - | 3 - 4 | 134 | 97 | ||
| KC692918.1 | - | 9 - 10 | 991 | 97 | ||
| L33412.1 | - | 9 - 10 | 999 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ager
Genomic Map