Assay Details
| Gene Symbol: Angpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007426.4 | NP_031452.2 | 5 - 6 | 1216 | 56 | |
| GenBank mRNA | AF004326.1 | - | 5 - 6 | 1142 | 56 | |
| AK019860.1 | - | 5 - 6 | 1205 | 56 | ||
| AK048622.1 | - | 5 - 6 | 1173 | 56 | ||
| AK143974.1 | - | 5 - 6 | 1204 | 56 | ||
| AK156132.1 | - | 5 - 6 | 1187 | 56 | ||
| BC027216.1 | - | 5 - 6 | 1169 | 3494566 | 56 | |
| BC138309.1 | - | 5 - 6 | 1110 | 8861329 | 56 | |
| BC138313.1 | - | 5 - 6 | 1110 | 8861333 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Angpt2
Genomic Map