Assay Details
| Gene Symbol: Apc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007462.3 | NP_031488.2 | 6 - 7 | 955 | 73 | |
| GenBank mRNA | AK045053.1 | - | 6 - 7 | 926 | 73 | |
| AK080907.1 | - | 6 - 7 | 955 | 73 | ||
| AK082724.1 | - | 6 - 7 | 701 | 73 | ||
| BC141141.1 | - | 5 - 6 | 668 | 9055753 | 73 | |
| M88127.1 | - | 5 - 6 | 644 | 73 | ||
More Information
Set Membership: |
|

Apc
Genomic Map