Assay Details
| Gene Symbol: Apc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007462.3 | NP_031488.2 | 8 - 9 | 1134 | 78 | |
| GenBank mRNA | AK045053.1 | - | 8 - 9 | 1105 | 78 | |
| AK046280.1 | - | 8 - 9 | 1124 | 78 | ||
| AK080907.1 | - | 8 - 9 | 1134 | 78 | ||
| BC141141.1 | - | 7 - 8 | 847 | 9055753 | 78 | |
| M88127.1 | - | 7 - 8 | 823 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Apc
Genomic Map