Assay Details
| Gene Symbol: Aplp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007467.3 | NP_031493.2 | 9 - 10 | 1341 | 63 | |
| GenBank mRNA | AK154992.1 | - | 9 - 10 | 1295 | 63 | |
| AK160871.1 | - | 9 - 10 | 1293 | 63 | ||
| AK170818.1 | - | 9 - 10 | 1274 | 63 | ||
| BC021877.1 | - | 9 - 10 | 1320 | 4481838 | 63 | |
| BC106158.1 | - | 8 - 9 | 1429 | 63 | ||
| L04538.1 | - | 9 - 10 | 1311 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aplp1
Genomic Map